Nalidixic acid 99.5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Ethyl-7-methyl-4-oxo-1,4-dihydro-1,8-naphthyridine-3-carboxylic acid

16990-1000 16990-0050 16990-0250
TS16990-0050EA 31 USD
TS16990-0050 TS16990-0250 TS16990-1000
Nalidixic acid 99.5%
Nalidixic acid
Formula: C₁₂H₁₂N₂O₃
MW: 232.24
Melting Pt: 225…231 °C
Storage Temperature: Ambient
MDL Number: MFCD00006884
CAS Number: 389-08-2
EINECS: 206-864-7

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 225 to 231 °C
Titration with NaOH ≥99.4 %
Specific optical rotation -44.0° to -48.0° (20 °C, 589 nm) (c=0.7, water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR