Cyclohexyl isocyanide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
15840-0050
15840-0250
15840-0010
TS15840-0250EA
411
USD
TS15840-0250
TS15840-0010
TS15840-0050
Cyclohexyl isocyanide 99%
Cyclohexyl isocyanide
Formula:
C₇H₁₁N Boiling Pt: 173…176 °C (760 mmHg) Density: 0.89 g/cm³ Flash Pt: 77 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00003839 CAS Number: 931-53-3 EINECS: 213-238-7 UN: 2810 ADR: 6.1,III |
Specification Test Results
Appearance | Clear colorless to light brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4490 to 1.4520 (20 °C, 589 nm) |
GC | ≥98.5 % |
UV | at 230 nm A: ≤0.15 |
UV | at 240 nm A: ≤0.12 |
UV | at 250 nm A: ≤0.11 |
UV | at 310 nm A: ≤0.05 |
UV | at 500 nm A: ≤0.02 |
pH | 8.5 to 9.6 (10 % in water) |
Titration with HClO4 | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...