Benzyl isocyanide 98%

Supplier: Thermo Scientific Chemicals
Warning

15839-0010 15839-0050 15839-0250
TS15839-0010EA 96 USD
TS15839-0010 TS15839-0050 TS15839-0250
Benzyl isocyanide 98%
Benzyl isocyanide
Formula: C₈H₇N
Boiling Pt: 105…106 °C (75 mmHg)
Density: 0.962 g/cm³
Flash Pt: 79 °C
Storage Temperature: Freezer
MDL Number: MFCD00000004
CAS Number: 10340-91-7
EINECS: 233-738-9
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Clear colorless to dark yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.5200 to 1.5220 (20 °C, 589 nm)
GC ≥97.5 %
Iron (Fe) ≤0.003 %
Potassium (K) ≤0.02 %
Heavy metals (as Pb) ≤0.003 %
Insoluble matter ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR