Luminol (3-Aminophthalhydrazide) 98%, pure

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Amino-2,3-dihydrophthalazine-1,4-dioneLuminolNSC 50645-Amino-2,3-Dihydro-1,4-Phthalazinedione

15385-0050 15385-0250 15385-1000 15385-0010
TS15385-0250EA 232 USD
TS15385-0250 TS15385-1000 TS15385-0010 TS15385-0050
Luminol (3-Aminophthalhydrazide) 98%, pure
Luminol (3-Aminophthalhydrazide)
Formula: C₈H₇N₃O₂
MW: 177.16 g/mol
Melting Pt: 319…320 °C
Storage Temperature: Ambient
MDL Number: MFCD00006890
CAS Number: 521-31-3
EINECS: 208-309-4

Order Now

Specification Test Results

Appearance Yellow to yellow-green to beige Powder and/or chunks
Infrared spectrum Conforms
Solubility (2 % in 1 N NaOH) Clear yellow to yellow-green
Titration excess NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR