2,2,6,6-Tetramethylpiperidin-4-ol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2,6,6-Tetramethyl-4-piperidinol, 4-Hydroxy-2,2,6,6-tetramethylpiperidine

15194-2500 15194-0050 15194-0250
TS15194-2500EA 242 USD
TS15194-2500 TS15194-0050 TS15194-0250
2,2,6,6-Tetramethylpiperidin-4-ol 99%
2,2,6,6-Tetramethylpiperidin-4-ol
Formula: C₉H₁₉NO
Melting Pt: 129…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00005983
CAS Number: 2403-88-5
EINECS: 219-291-2

Order Now

Specification Test Results

Appearance White to off-white to light beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 129 to 133 °C
Titration with HClO4 ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR