(S)-(-)-2-Pyrrolidinone-5-carboxylic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
13212-0250
13212-1000
13212-5000
TS13212-0250EA
31
USD
TS13212-0250
TS13212-1000
TS13212-5000
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid 98%
(S)-(-)-2-Pyrrolidinone-5-carboxylic acid
Formula:
C₅H₇NO₃ Melting Pt: 152…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005272 CAS Number: 98-79-3 EINECS: 202-700-3 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤0.5 % (K.F.) |
Specific optical rotation | -26° to -29° (20 °C, 589 nm) (c=10, 1N NaOH) (on dry substance) |
Titration with NaOH | ≥97.5 % (On dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...