N,O-Dimethylhydroxylamine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
11634-0050
11634-0250
11634-1000
TS11634-0050EA
52
USD
TS11634-0050
TS11634-0250
TS11634-1000
N,O-Dimethylhydroxylamine hydrochloride 98%
N,O-Dimethylhydroxylamine hydrochloride
Formula:
C₂H₈ClNO Melting Pt: 112…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012485 CAS Number: 6638-79-5 EINECS: 229-642-1 |
Specification Test Results
Appearance | White to off-white Crystalline powder or crystals |
Infrared spectrum | Conforms |
Melting point | 112 to 116 °C |
Water | ≤0.5 % |
Solubility | (10 % in water) Clear |
Titration with HClO4 | ≥97.5 % (on anhydrous substance) |
Sulfate (SO4) | ≤0.003 % |
Iron (Fe) | ≤5 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...