N,N-Dimethyl-1,3-diaminopropane 99%

Supplier: Thermo Scientific Chemicals
Danger

11590-0010 11590-0025 11590-0050 11590-2500
TS11590-0010EA 48 USD
TS11590-0010 TS11590-0025 TS11590-0050 TS11590-2500
N,N-Dimethyl-1,3-diaminopropane 99%
N,N-Dimethyl-1,3-diaminopropane
Formula: C₅H₁₄N₂
Boiling Pt: 133 °C (760 mmHg)
Melting Pt: –60 °C
Density: 0.81 g/cm³
Flash Pt: 32 °C
Storage Temperature: Ambient
MDL Number: MFCD00008216
CAS Number: 109-55-7
EINECS: 203-680-9
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4340 to 1.4360 (20 °C, 589 nm)
Water ≤0.5 %
GC ≥98.5 %
Color scale ≤15 APHA
Insoluble matter ≤1 % (in acetic acid)
NH4OH precipitate ≤2.0 %
Loss on ignition ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR