α-Benzoinoxime 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: α-Benzoin oxime2-Hydroxy-1,2-diphenylethanone oxime

10553-5000 10553-0250 10553-1000
TS10553-0250EA 45.8 USD
TS10553-0250 TS10553-1000 TS10553-5000
α-Benzoinoxime 98%
α-Benzoinoxime
Formula: C₁₄H₁₃NO₂
MW: 227.26 g/mol
Melting Pt: 150…155 °C
Storage Temperature: Ambient
MDL Number: MFCD00004501
CAS Number: 441-38-3
EINECS: 207-127-2

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 155 °C
HPLC ≥97.5 %
GC ≥99.0 %
UV at 210 nm A: ≤1
UV at 220 nm A: ≤0.35
UV at 235 nm A: ≤0.16
UV at 240 nm A: ≤0.05
UV at 250 nm A: ≤0.01
UV at 300 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Substance darkened by H2SO4 passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR