(S)-(-)-1-Phenylethyl isocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (S)-1-phenylethylisocyanate(S)-(-)-α-Phenethyl isocyanate(S)-(-)-α-Methylbenzyl isocyanate(S)-(-)-1-Phenethyl isocyanate

20531-0010 20531-0050
TS20531-0010EA 63 USD
TS20531-0010 TS20531-0050
(S)-(-)-1-Phenylethyl isocyanate 98%
(S)-(-)-1-Phenylethyl isocyanate
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 55…56 °C (2.5 mmHg)
Density: 1.05 g/cm³
Flash Pt: 65 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002036
CAS Number: 14649-03-7
EINECS: 238-698-6
UN: 2206
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5130 to 1.5150 (20 °C, 589 nm)
GC ≥97.5 %
Specific optical rotation -9° to -11° (20 °C, 589 nm) (neat)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR