(-)-10-Camphorsulfonic acid 98%

Supplier: Thermo Scientific Chemicals
Danger

26364-1000 26364-5000 26364-0050 26364-0250
TS26364-1000EA 103 USD
TS26364-1000 TS26364-5000 TS26364-0050 TS26364-0250
(-)-10-Camphorsulfonic acid 98%
L(-)-Camphorsulfonic acid
Formula: C₁₀H₁₆O₄S
Boiling Pt: 375 °C (1013 hPa)
Melting Pt: 198 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064158
CAS Number: 35963-20-3
EINECS: 252-817-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Specific optical rotation -21° to -23° (20°C, 589 nm) (c=20, H2O)
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR