Norethindrone

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Norethisterone

26289-0010 26289-0050
TS26289-0010EA 104 USD
TS26289-0010 TS26289-0050
Norethindrone
Norethindrone
Formula: C₂₀H₂₆O₂
Melting Pt: 205…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00067596
CAS Number: 68-22-4

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 205 to 208 °C
Loss on drying ≤0.5 % (1 g, 105 °C, 3 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR