Dipropylamine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Propyl-1-propanamine

11749-2500 11749-0010
TS11749-2500EA 21.9 USD
TS11749-2500 TS11749-0010
Dipropylamine 99%
Dipropylamine
Formula: C₆H₁₅N
MW: 101.19 g/mol
Boiling Pt: 109.4 °C (1013 hPa)
Melting Pt: –63 °C
Density: 0.74 g/cm³
Flash Pt: 7 °C
Storage Temperature: Ambient
MDL Number: MFCD00009362
CAS Number: 142-84-7
EINECS: 205-565-9
UN: 2383
ADR: 3,II
Merck Index: 12,03406

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4030 to 1.4050 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %
Color scale ≤15 APHA
Heavy metals (ICP-OES) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR