N,N-Dimethyl-3-nitroaniline 98%

Supplier: Thermo Scientific Chemicals
Warning

11647-0250 11647-0050
TS11647-0250EA 91 USD
TS11647-0250 TS11647-0050
N,N-Dimethyl-3-nitroaniline 98%
N,N-Dimethyl-3-nitroaniline
Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
Boiling Pt: 280…285 °C (1013 hPa)
Melting Pt: 57…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00007236
CAS Number: 619-31-8
EINECS: 210-590-3
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Orange to orange-red Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 57 to 61 °C
Titration with HClO4 ≥97.5 %
Nitrate (NO3) ≤0.001 %
Sulfate (SO4) ≤0.01 %
Calcium (Ca) ≤0.001 %
Iron (Fe) ≤0.001 %
Magnesium (Mg) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR