3-(Dimethylamino)phenol 97%

Supplier: Thermo Scientific Chemicals
Warning

11586-0250 11586-5000
TS11586-0250EA 110 USD
TS11586-0250 TS11586-5000
3-(Dimethylamino)phenol 97%
3-(Dimethylamino)phenol
Formula: C₈H₁₁NO
Melting Pt: 80…85 °C
Flash Pt: 148 °C
Storage Temperature: Ambient
MDL Number: MFCD00002264
CAS Number: 99-07-0
EINECS: 202-727-0

Order Now

Specification Test Results

Appearance Grey to brown to purple Powder
Infrared spectrum Conforms
GC Xylene isomers plus Ethylbenzene (Ethylbenzene not to exceed 25%)
Water ≤0.05 % (Coulometric)
Color scale ≤10 APHA
Residue after evaporation ≤0.002 %
Substance darkened by H2SO4 Passes test
Sulfur compounds (as S) ≤0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR