4-(Dimethylamino)antipyrine 97%

Supplier: Thermo Scientific Chemicals
Warning

11574-0010 11574-0250 11574-1000 11574-0050
TS11574-0010EA 564 USD
TS11574-0010 TS11574-1000 TS11574-0050 TS11574-0250
4-(Dimethylamino)antipyrine 97%
4-(Dimethylamino)antipyrine
Formula: C₁₃H₁₇N₃O
MW: 231.3 g/mol
Melting Pt: 105…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00003142
CAS Number: 58-15-1
EINECS: 200-365-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder or crystals
Infrared spectrum Conforms
Melting point 105 to 110 °C
Titration with HClO4 ≥96.0 %
Specific optical rotation -15° to -18° (20 °C, 589 nm) (c=10, EtOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR