Diethylenetriamine 98+%

Supplier: Thermo Scientific Chemicals
Danger

11431-0025 11431-0100 11431-2500
TS11431-0010EA 79 USD
TS11431-0010 TS11431-0025 TS11431-0100 TS11431-2500
Diethylenetriamine 98+%
Diethylenetriamine
Formula: C₄H₁₃N₃
Boiling Pt: 206 °C (760 mmHg)
Melting Pt: –40 °C
Density: 0.96 g/cm³
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00008171
CAS Number: 111-40-0
EINECS: 203-865-4
UN: 2079
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4820 to 1.4850 (20°C, 589 nm)
GC ≥98.0 %
Color scale ≤50 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR