Adiponitrile 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexanedinitrile

11383-0010 11383-1000
TS11383-0010EA 62 USD
TS11383-0010 TS11383-1000
Adiponitrile 99%
Adiponitrile
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 296
Melting Pt: 2
Density: 0.95 g/cm³
Flash Pt: 163 °C
MDL Number: MFCD00001975
CAS Number: 111-69-3
EINECS: 203-896-3
UN: 2205
ADR: 6.1 III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4370 to 1.4390 (20°C, 589 nm)
GC ≥98.5 %
Microbial contamination (Salmonella) Absent in 25 g
pH 6 to 8 (50 % suspension in water)
Sulfite (as SO2) ≤10 ppm
Arsenic (As) ≤0.07 ppm
Cadmium (Cd) ≤0.05 ppm
Iron (Fe) ≤10 ppm
Lead (Pb) ≤0.1 ppm
Mercury (Hg) ≤0.05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR