Terephthalonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,4-Benzenedicarbonitrile, Terephthalodinitrile, Benzene-1,4-dicarbonitrile, 1,4-Dicyanobenzene

11382-0250 11382-1000 11382-5000
TS11382-0250EA 34 USD
TS11382-0250 TS11382-1000 TS11382-5000
Terephthalonitrile 98%
Terephthalonitrile
Formula: C₈H₄N₂
Melting Pt: 222…227 °C
Storage Temperature: Ambient
MDL Number: MFCD00001810
CAS Number: 623-26-7
EINECS: 210-783-2

Order Now

Specification Test Results

Appearance White to light yellow to light beige Powder
Infrared spectrum Conforms
Melting point 223 to 230 °C
HPLC ≥97.5 %
Ammonium (NH4) ≤20 ppm
Iron (Fe) ≤2 ppm
Sulfur (S) ≤10 ppm
Heavy metals (as Pb) ≤5 ppm
Impurity ≤0.2 % Carbonate (as Na2CO3)
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR