Cyanoguanidine 99.5%

Supplier: Thermo Scientific Chemicals

11381-0010 11381-0025 11381-0050 11381-0250
TS11381-0010EA 48 USD
TS11381-0010 TS11381-0025 TS11381-0050 TS11381-0250
Cyanoguanidine 99.5%
Cyanoguanidine
Formula: (H₂N)₂CNCN
Melting Pt: 208…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00008066
CAS Number: 461-58-5
EINECS: 207-312-8
Merck Index: 13,03119

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 208 to 211 °C
Assay ≥99.4 % (ex N)
Water ≤0.5 % (K.F.)
Sulfated ash ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR