1,3-Diamino-2-propanol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Diaminopropan-2-ol1,3-Diamino-2-hydroxypropane

11222-0250 11222-0050
TS11222-0050EA 52 USD
TS11222-0050 TS11222-0250
1,3-Diamino-2-propanol 95%
1,3-Diamino-2-propanol
Formula: C₃H₁₀N₂O
MW: 90.13 g/mol
Boiling Pt: 114 °C (5 mmHg)
Melting Pt: 39…45 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008142
CAS Number: 616-29-5
EINECS: 210-474-2

Order Now

Specification Test Results

Appearance White to yellow Low melting solid
Infrared spectrum Conforms
Titration with HClO4 ≥94 %
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR