Carbachol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Carbamylcholine chlorideCarbamoylcholine chloride

10824-0050 10824-0250
TS10824-0050EA 102 USD
TS10824-0050 TS10824-0250
Carbachol 99%
Carbachol
Formula: H₂NC(O)O(CH₂)₂N(CH₃)₃Cl
MW: 182.65 g/mol
Melting Pt: 200…204 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012011
CAS Number: 51-83-2
EINECS: 200-127-3
UN: 2811
ADR: 6.1,II
Merck Index: 13,01786

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 200 to 204 °C (Dec)
Loss on drying ≤2.0 % (105 °C, 2 hrs)
Titration with HClO4 ≥98.5 % (On dried substance)
UV at 240 nm A: ≤0.04
UV at 230 nm A: ≤0.10
UV at 220 nm A: ≤0.20
UV at 210 nm A: ≤0.30
UV at 195 nm A: ≤1.0
Color scale ≤10 APHA
Residue after evaporation ≤0.001 %
Sulfur compounds (as S) ≤0.005 %
Water soluble titr. acid ≤0.0003 meq/g
Thiophene passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR