(±)-sec-Butylamine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
(±)-2-Aminobutane
, (±)-2-Butylamine
10781-0050
10781-5000
10781-0025
10781-1000
10781-0250
TS10781-0025EA
170
USD
TS10781-0025
TS10781-1000
TS10781-5000
TS10781-0250
TS10781-0050
(±)-sec-Butylamine 99%
(±)-sec-Butylamine
Formula:
C₄H₁₁N MW: 73.14 g/mol Boiling Pt: 63 °C (760 mmHg) Melting Pt: –104 °C Density: 0.72 g/cm³ Flash Pt: –19 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008094 CAS Number: 13952-84-6 EINECS: 237-732-7 UN: 2734 ADR: 8,I |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3920 to 1.3960 (20°C, 589 nm) |
GC | ≥98.5 % |
Titration with NaOH | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...