2-(2-Aminoethyl)pyridine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Pyridylethylamine, 2-(2-Pyridyl)ethylamine, 2-Pyridin-2-ylethanamine

10369-0010 10369-0100 10369-0500
TS10369-0010EA 33 USD
TS10369-0010 TS10369-0100 TS10369-0500
2-(2-Aminoethyl)pyridine 95%
2-(2-Aminoethyl)pyridine
Formula: C₇H₁₀N₂
MW: 122.17 g/mol
Boiling Pt: 105…106 °C (10 mmHg)
Density: 1.021 g/cm³
Flash Pt: 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00006367
CAS Number: 2706-56-1
EINECS: 220-295-1
UN: 2735

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5340 to 1.5380 (20°C, 589 nm)
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR