1-Methylimidazole (NMI), anhydrous, AcroSeal™ for biochemistry

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI

46529-0010 465290025
TS46529-0010EA 378 USD
TS46529-0010 TS465290025
1-Methylimidazole (NMI), anhydrous, AcroSeal™ for biochemistry
1-Methylimidazole
Formula: C₄H₆N₂
MW: 82.11 g/mol
Boiling Pt: 198 °C
Melting Pt: –2 °C
Density: 1.03 g/cm³
Flash Pt: 92 °C
MDL Number: MFCD00005292
CAS Number: 616-47-7
UN: 2922
ADR: 8 II

Order Now

Specification Test Results

Appearance Clear colorless Liquid (May discolor to yellow on storage)
Infrared spectrum Conforms
Refractive index 1.4955 to 1.4975 (20°C, 589 nm)
Water ≤0.005 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR