Tenofovir disoproxil fumarate 98%

Supplier: Thermo Scientific Chemicals

46125-0010 46125-0050 46125-0250
TS46125-0010EA 49 USD
TS46125-0010 TS46125-0050 TS46125-0250
Tenofovir disoproxil fumarate 98%
Tenofovir disoproxil fumarate
Formula: C₁₉H₃₀N₅O₁₀P·C₄H₄O₄
Storage Temperature: Refrigerator
MDL Number: MFCD08141829
CAS Number: 202138-50-9

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1 %
HPLC ≥97.5 %
Fumaric acid 17.5 to 19


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR