Ritonavir 98%

Supplier: Thermo Scientific Chemicals
Warning

46122-0010 46122-2500
TS46122-0010EA 494 USD
TS46122-0010 TS46122-2500
Ritonavir 98%
Ritonavir
Formula: C₃₇H₄₈N₆O₅S₂
Melting Pt: 120…132 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00927142
CAS Number: 155213-67-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 121 to 127 °C
Water ≤0.5 %
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR