Pyrimethamine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-(4-Chlorophenyl)-6-ethylpyrimidine-2,4-diamine, 5-(4-Chlorophenyl)-6-ethyl-2,4-pyrimidinediamine

46120-0010 46120-0050
TS46120-0010EA 35 USD
TS46120-0010 TS46120-0050
Pyrimethamine 98%
Pyrimethamine
Formula: C₁₂H₁₃ClN₄
MW: 248.71 g/mol
Melting Pt: 241 °C
Storage Temperature: Ambient
MDL Number: MFCD00057350
CAS Number: 58-14-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 239 to 243 °C
Loss on drying ≤0.5 % (105 °C, 4 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR