Azaperone

Supplier: Thermo Scientific Chemicals
Danger

46083-0010 46083-0050 460830250
TS46083-0010EA 102 USD
TS46083-0010 TS46083-0050 TS460830250
Azaperone
Azaperone
Formula: C₁₉H₂₂FN₃₀
Storage Temperature: Refrigerator
MDL Number: MFCD00866692
CAS Number: 1649-18-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 92 to 95 °C
Loss on drying ≤0.5 % (60 °C, 4 h) (vacuum)
Titration with HClO4 99 to 101 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR