Ribavirin 98%

Supplier: Thermo Scientific Chemicals
Danger

46048-0010 46048-0050 46048-0250
TS46048-0010EA 105 USD
TS46048-0010 TS46048-0050 TS46048-0250
Ribavirin 98%
Ribavirin
Formula: C8H12N4O5
Storage Temperature: Refrigerator
MDL Number: MFCD00058564
CAS Number: 36791-04-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 %
HPLC ≥97.5 %
Residue after ign. (sulfated) ≤0.25 %
Specific optical rotation -33° to -37° (20°C, 589 nm) (1 % in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR