Risperidone 99%

Supplier: Thermo Scientific Chemicals
Danger

45939-0050 459390010
TS45939-0010EA 75 USD
TS45939-0010 TS45939-0050 TS459390010
Risperidone 99%
Risperidone

Soluble in chloroform, methanol.

Formula: C₂₃H₂₇FN₄O₂
Melting Pt: 172 °C
Storage Temperature: Ambient
MDL Number: MFCD00274576
CAS Number: 106266-06-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 170 to 174 °C
Loss on drying ≤0.5 %
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR