Aripiprazole

Supplier: Thermo Scientific Chemicals
Warning

45799-0010 45799-0050
TS45799-0010EA 92 USD
TS45799-0010 TS45799-0050
Aripiprazole
Aripiprazole
Formula: C₂₃H₂₇Cl₂N₃O₂
Melting Pt: 149 °C
Storage Temperature: Ambient
MDL Number: MFCD00892072
CAS Number: 129722-12-9

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Melting point 133 to 149 °C
Loss on drying ≤0.3 % (105 °C, 3 h)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR