Choline hydrogen tartrate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-N,N,N-trimethylethan-1-aminium (2R,3R)-3-carboxy-2,3-dihydroxypropanoate, Choline acid tartrate, CBT, 2-(Hydroxyethyl)trimethylammonium bitartrate, Choline bitartrate

45022-1000 45022-5000
TS45022-1000EA 48 USD
TS45022-1000 TS45022-5000
Choline hydrogen tartrate 97%
Choline hydrogen tartrate
Formula: C₉H₁₉NO₇
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00036332
CAS Number: 87-67-2

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 %
Titration with HClO4 ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR