6-O-Palmitoyl-L-ascorbic acid, meets analytical specification of Ph. Eur., USP

Supplier: Thermo Scientific Chemicals

Synonyms: L-Ascorbic acid 6-palmitate, Palmitoyl-L-ascorbic acid, Ascorbyl Palmitate, (S)-2-((R)-3,4-Dihydroxy-5-oxo-2,5-dihydrofuran-2-yl)-2-hydroxyethyl palmitate, L(+)-Ascorbyl palmitate

44948-0250 44948-1000
TS44948-0250EA 58 USD
TS44948-0250 TS44948-1000
6-O-Palmitoyl-L-ascorbic acid, meets analytical specification of Ph. Eur., USP
6-O-Palmitoyl-L-ascorbic acid
Formula: C₂₂H₃₈O₇
MW: 414.54 g/mol
Boiling Pt: 250 °C
Melting Pt: 111…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00005377
CAS Number: 137-66-6

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 107 to 117 °C
Titration Iodimetric 98.0 to 100.5 % (on dry substance)
Loss on drying ≤1 % (60 °C, 5 Hrs) (vacuum)
Heavy metals ≤10 ppm
Appearance of solution Passes test
Residue after ignition ≤0.1 %
Specific optical rotation +21° to +24° (20 °C, 589 nm) (c=10, methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR