Sodium sulfate, anhydrous 99% for biochemistry

Supplier: Thermo Scientific Chemicals

44792-0025 44792-5000
TS44792-0025EA 228 USD
TS44792-0025 TS44792-5000
Sodium sulfate, anhydrous 99% for biochemistry
Sodium sulphate
Formula: Na₂SO₄
Boiling Pt: >1700 °C (760 mmHg)
Melting Pt: 884 °C
Storage Temperature: Ambient
MDL Number: MFCD00003504
CAS Number: 7757-82-6

Order Now

Specification Test Results

Appearance White Crystalline powder
Loss on drying ≤0.5 %
Solubility (10 % in water) Clear colorless
Acidity/alkalinity Passes test
pH 5.0 to 9.2
Titration with HClO4 ≥98.5 %
Chloride (Cl) ≤0.001 %
Phosphate (PO4) ≤0.001 %
Arsenic (As) ≤0.0001 %
Calcium (Ca) ≤0.005 %
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Potassium (K) ≤0.002 %
Zinc (Zn) ≤0.002 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR