Sodium azide 99+% for biochemistry

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Sodium triazide

44781-5000 44781-0250 44781-1000
TS44781-0250EA 82.6 USD
TS44781-0250 TS44781-1000 TS44781-5000
Sodium azide 99+% for biochemistry
Sodium azide
Formula: NaN₃
MW: 65.01 g/mol
Boiling Pt: 300 °C (760 mmHg)
Melting Pt: 275 °C (dec.)
Density: 1.85 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
UN: 1687
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to off-white Powder
Titration Argentometric ≥99.0 %
UV (1 M in water)
UV at 275 nm Cut off
Heavy metals (as Pb) ≤10 ppm
Insoluble matter ≤0.05 % (in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR