Magnesium sulfate heptahydrate 99% for biochemistry
Supplier: Thermo Scientific Chemicals
Synonyms:
sulfuric acid magnesium salt heptahydrate
, Bitter salt
, sulphuric acid magnesium salt heptahydrate
, Epsom salt
44716-5000
44716-0025
TS44716-5000EA
106
USD
TS44716-5000
TS44716-0025
Magnesium sulfate heptahydrate 99% for biochemistry
Magnesium sulphate heptahydrate
Formula:
MgSO₄·7H₂O MW: 246.48 g/mol Melting Pt: 770 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149785 CAS Number: 10034-99-8 |
Specification Test Results
Appearance | White Crystals |
Titration Complexometric | ≥98.5 % |
Solubility | (1 M in water) Clear colorless |
pH | 5 to 8 |
Heavy metals (as Pb) | ≤5 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...