Ammonium acetate 98% for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: Acetic acid ammonium salt

44712-0025 44712-1000 44712-5000
TS44712-0025EA 289 USD
TS44712-0025 TS44712-1000 TS44712-5000
Ammonium acetate 98% for biochemistry
Ammonium acetate
Formula: H₃CCOONH₄
Melting Pt: 114 °C
Flash Pt: 136 °C
Storage Temperature: Ambient
MDL Number: MFCD00013066
CAS Number: 631-61-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Solubility (1 M in water) Clear colorless
UV at 350 nm A: ≤0.01
pH 6.5 to 7.3 (5 % in water)
Titration with HClO4 ≥97.5 %
Heavy metals (as Pb) ≤2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR