Dextran sulfate sodium salt, M.W. ∼ 500,000
Supplier: Thermo Scientific Chemicals
Synonyms:
Dextran sulfate sodium from Dextran
, Dextran sulphate sodium from Dextran
43324-0250
43324-1000
43324-0050
TS43324-1000EA
354.14
USD
TS43324-1000
TS43324-0050
TS43324-0250
Dextran sulfate sodium salt, M.W. ∼ 500,000
Dextran sulphate sodium salt
Formula:
(C₆H₇Na₃O₁₄S₃)n Storage Temperature: Refrigerator |
MDL Number:
MFCD00081551 CAS Number: 9011-18-1 |
Specification Test Results
Appearance | Off-white to light yellow Powder |
Loss on drying | ≤10 % |
Sulfated ash | 40 % to 50 % |
Solubility | (10 % in water) Clear to slightly hazy colorless to light yellow |
Molecular weight | ≥500000 |
pH | 5 to 7.5 (1 % in water) |
Specific optical rotation | +75° to +105° (20°C, 589 nm) (c=6, water) |
Specific viscosity | 0.10 to 0.70 (1.0M in NaCl, 25°C) |
Sulfur (S) | 17 to 20 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...