oxolinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

43151-0050 43151-0250
TS43151-0050EA 113 USD
TS43151-0050 TS43151-0250
oxolinic acid 98%
oxolinic acid
Formula: C₁₃H₁₁NO₅
MW: 261.233 g/mol
Melting Pt: 314…316 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00056775
CAS Number: 14698-29-4

Order Now

Specification Test Results

Appearance White to almost white Crystalline powder
Infrared spectrum Conforms
Melting point 311 to 314 °C (Dec)
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR