Fmoc-Cys(tBu)-OH 98%

Supplier: Thermo Scientific Chemicals

43127-0250 43127-0050
TS43127-0250EA 270 USD
TS43127-0250 TS43127-0050
Fmoc-Cys(tBu)-OH 98%
Fmoc-Cys(tBu)-OH
Formula: C₂₂H₂₅NO₄S
Melting Pt: 137.1…138.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00037130
CAS Number: 67436-13-9

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
HPLC ≥97.5 %
Specific optical rotation -21.0° to -23.0° (20 °C, 589 nm) (c=1, DMF)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR