4-Oxazolecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Oxazole-4-carboxylic acid
43011-0050
43011-0010
TS43011-0050EA
384
USD
TS43011-0050
TS43011-0010
4-Oxazolecarboxylic acid 97%
4-Oxazolecarboxylic acid
Formula:
C₄H₃NO₃ MW: 113.0715 g/mol Melting Pt: 141…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD06797151 CAS Number: 23012-13-7 |
Specification Test Results
Appearance | Light orange to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
GC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...