2-Amino-5-thiazolecarboxylic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Aminothiazole-5-carboxylic acid
42957-0010
42957-0050
TS42957-0010EA
196
USD
TS42957-0010
TS42957-0050
2-Amino-5-thiazolecarboxylic acid 95%
2-Amino-5-thiazolecarboxylic acid
Formula:
C₄H₄N₂O₂S Melting Pt: 182…184 °C Storage Temperature: Ambient |
MDL Number:
MFCD06203554 CAS Number: 40283-46-3 |
Specification Test Results
Appearance | Off-white to yellow Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Color scale | ≤10 APHA |
Non-volatile matter | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...