Diphenylamine ACS

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-PhenylbenzeneamineN-Phenylaniline

42365-0050 42365-5000 42365-1000
TS42365-1000EA 79.14 USD
TS42365-1000 TS42365-0050 TS42365-5000
Diphenylamine ACS
Diphenylamine
Formula: (C₆H₅)₂NH
MW: 169.23 g/mol
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 52…54 °C
Flash Pt: 152 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00003014
CAS Number: 122-39-4
EINECS: 204-539-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals or flakes
Infrared spectrum Conforms
Melting point 52.5 to 54.0 °C
Solubility (in alcohol) Passes test
Residue after ignition ≤0.03 %
Sensitivity (to nitrate) Passes test
Nitrate (NO3) Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR