Wright's stain, pure, certified

Supplier: Thermo Scientific Chemicals
Warning

42264-0100 42264-0250 42264-1000
TS42264-0100EA 35 USD
TS42264-0100 TS42264-0250 TS42264-1000
Wright's stain, pure, certified
Wright's stain
Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1
EINECS: 273-541-5

Order Now

Specification Test Results

Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
BSC Certification Passes


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR