Potassium chloride for biochemistry

Supplier: Thermo Scientific Chemicals

Synonyms: Potassiumchloride

41820-0025 41820-5000
TS41820-0025EA 334 USD
TS41820-0025 TS41820-5000
Potassium chloride for biochemistry
Potassium chloride
Formula: KCl
Boiling Pt: 1420 °C (760 mmHg)
Melting Pt: 770 °C
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Titration Argentometric ≥99.0 %
Solubility (1 M in water) Clear colorless
UV at 280 nm A: ≤0.05
pH 5.5 to 8.0 (5 % in water)
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.001 %
Heavy metals (as Pb) ≤5 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR