Dextran for biochemistry, high fraction

Supplier: Thermo Scientific Chemicals

40626-0010 40626-1000
TS40626-0010EA 726 USD
TS40626-0010 TS40626-1000
Dextran for biochemistry, high fraction
Dextran
Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Loss on drying ≤9.0 %
Sulfated ash ≤2.0 %
UV (6 % in water)
UV at 375 nm A: ≤0.15
Viscosity 0.4 to 0.5 (1 % at 37°C)
Average molecular weight 250000
pH 4.5 to 7.0 (6 % in water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR