Methyl-4-(aminomethyl)benzoate hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(Aminomethyl)benzoic Acid Methyl Ester HydrochlorideMethyl 4-(aminomethyl)benzoate hydrochloride

39678-0250 39678-0010 39678-0050
TS39678-0050EA 72 USD
TS39678-0050 TS39678-0250 TS39678-0010
Methyl-4-(aminomethyl)benzoate hydrochloride 97%
Methyl-4-(aminomethyl)benzoate hydrochloride
Formula: C₉H₁₁NO₂
MW: 201.65 g/mol
Melting Pt: 234.5…237 °C
Storage Temperature: Ambient
MDL Number: MFCD00182671
CAS Number: 6232-11-7

Order Now

Specification Test Results

Appearance White Crystalline powder or crystals
Infrared spectrum Conforms
Titration Argentometric 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR