4-Amino-m-anisic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Amino-3-methoxybenzenecarboxylic acid

39146-0010 39146-0050
TS39146-0010EA 63 USD
TS39146-0010 TS39146-0050
4-Amino-m-anisic acid 98%
4-Amino-m-anisic acid
Formula: C₈H₉NO₃
MW: 167.16
Melting Pt: 186…188 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00016539
CAS Number: 2486-69-3

Order Now

Specification Test Results

Appearance White to light beige to grey Powder
Infrared spectrum Conforms
Melting point 186 to 191 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR