Acetamidoxime 95+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Acetamide oxime

39053-0010 39053-0050
TS39053-0010EA 28 USD
TS39053-0010 TS39053-0050
Acetamidoxime 95+%
Acetamidoxime
Formula: C₂H₆N₂O
MW: 74.08 g/mol
Melting Pt: 134…138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00603514
CAS Number: 22059-22-9

Order Now

Specification Test Results

Appearance White to off-white Crystals
Infrared spectrum Conforms
Melting point 131 to 138 °C
Titration with HClO4 ≥95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR